Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198423.3 | NP_940815.3 | 25 - 26 | 6690 | 64 | |
GenBank mRNA | AB257855.1 | - | 24 - 25 | 6692 | 64 | |
AK147250.1 | - | 25 - 26 | 6690 | 64 | ||
AK147400.1 | - | 17 - 18 | 3598 | 64 | ||
AK164229.1 | - | 8 - 9 | 931 | 64 | ||
BC060615.1 | - | 8 - 9 | 962 | 64 | ||
BC072602.1 | - | 14 - 15 | 2128 | 6830770 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |