Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026197.3 | NP_080473.1 | 6 - 7 | 846 | 99 | |
GenBank mRNA | AK011780.1 | - | 6 - 7 | 847 | 99 | |
AK012739.1 | - | 6 - 7 | 847 | 99 | ||
AK013306.1 | - | 6 - 7 | 858 | 99 | ||
AK029045.1 | - | 6 - 7 | 829 | 98 | ||
AK145022.1 | - | 6 - 7 | 854 | 99 | ||
AK145662.1 | - | 6 - 7 | 846 | 99 | ||
AK168798.1 | - | 6 - 7 | 796 | 99 | ||
BC003467.1 | - | 6 - 7 | 100 | |||
BC049897.1 | - | 6 - 7 | 775 | 3979938 | 99 | |
BC054724.1 | - | 6 - 7 | 782 | 5695738 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |