Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286200.1 | NP_001273129.1 | 6 - 7 | 992 | 83 | |
NM_009353.2 | NP_033379.2 | 6 - 7 | 992 | 83 | ||
NR_104410.1 | 7 - 8 | 1092 | 83 | |||
GenBank mRNA | AF003000.1 | - | 6 - 7 | 886 | 83 | |
AK089419.1 | - | 6 - 7 | 967 | 83 | ||
AK160421.1 | - | 7 - 8 | 737 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |