Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172630.2 | NP_766218.1 | 7 - 8 | 1316 | 85 | |
GenBank mRNA | AK041262.1 | - | 7 - 8 | 1317 | 85 | |
AK154265.1 | - | 7 - 8 | 1170 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |