Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013025.2 | NP_001013043.1 | 4 - 5 | 1447 | 104 | |
GenBank mRNA | AK141742.1 | - | 3 - 4 | 1003 | 104 | |
BC072608.1 | - | 4 - 5 | 1447 | 104 | ||
BC138866.1 | - | 4 - 5 | 1361 | 8861888 | 104 | |
BC138867.1 | - | 4 - 5 | 1361 | 8861889 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |