Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037726.1 | NP_001032815.1 | 6 - 7 | 942 | 112 | |
NM_009952.2 | NP_034082.1 | 8 - 9 | 1106 | 112 | ||
NM_133828.2 | NP_598589.2 | 7 - 8 | 1064 | 112 | ||
GenBank mRNA | AF448507.1 | - | 8 - 9 | 982 | 112 | |
AF448508.1 | - | 1634 | 112 | |||
AK044992.1 | - | 7 - 8 | 879 | 112 | ||
AK044993.1 | - | 7 - 8 | 879 | 112 | ||
BC021649.1 | - | 7 - 8 | 997 | 4017170 | 112 | |
M95106.1 | - | 8 - 9 | 1016 | 112 | ||
U46027.1 | - | 5 - 6 | 629 | 112 | ||
X92497.1 | - | 6 - 7 | 919 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |