Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172422.3 | NP_766010.1 | 6 - 7 | 1361 | 135 | |
GenBank mRNA | AK035322.1 | - | 6 - 7 | 1361 | 135 | |
AK084971.1 | - | 6 - 7 | 1336 | 135 | ||
AK129251.1 | - | 4 - 5 | 2538 | 135 | ||
AK148224.1 | - | 1345 | 135 | |||
AK153916.1 | - | 5 - 6 | 1304 | 135 | ||
AK166378.1 | - | 6 - 7 | 1346 | 135 | ||
AK167714.1 | - | 5 - 6 | 1617 | 135 | ||
BC008271.1 | - | 5 - 6 | 1302 | 3603187 | 135 | |
BC057208.1 | - | 5 - 6 | 1295 | 5328766 | 135 | |
BC080813.1 | - | 5 - 6 | 1301 | 6394260 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |