Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002238.2 | NP_001002238.1 | 24 - 25 | 3513 | 148 | |
NM_001002240.2 | NP_001002240.1 | 24 - 25 | 3486 | 148 | ||
NM_001002241.2 | NP_001002241.1 | 23 - 24 | 3786 | 148 | ||
NM_031387.3 | NP_113564.2 | 24 - 25 | 3646 | 148 | ||
GenBank mRNA | AB067571.1 | - | 23 - 24 | 3467 | 148 | |
AB183526.1 | - | 24 - 25 | 3513 | 148 | ||
AB183527.1 | - | 24 - 25 | 3486 | 148 | ||
AB183528.1 | - | 23 - 24 | 3786 | 148 | ||
AB183529.1 | - | 23 - 24 | 3350 | 148 | ||
AF285591.1 | - | 20 - 21 | 2840 | 148 | ||
AK029806.1 | - | 24 - 25 | 3647 | 148 | ||
BC129954.1 | - | 19 - 20 | 2772 | 40081321 | 148 | |
BC129955.1 | - | 19 - 20 | 2772 | 40081323 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |