Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103177.2 | NP_001096647.1 | 18 - 19 | 2088 | 74 | |
NM_001103178.2 | NP_001096648.1 | 16 - 17 | 1630 | 74 | ||
NM_001290813.1 | NP_001277742.1 | 17 - 18 | 1933 | 74 | ||
NM_001290815.1 | NP_001277744.1 | 17 - 18 | 1947 | 74 | ||
NM_001290816.1 | NP_001277745.1 | 17 - 18 | 1989 | 74 | ||
NM_178688.3 | NP_848803.3 | 21 - 22 | 2326 | 74 | ||
GenBank mRNA | AF404774.1 | - | 21 - 22 | 2326 | 74 | |
AF404775.1 | - | 18 - 19 | 1848 | 74 | ||
AF404776.1 | - | 16 - 17 | 1590 | 74 | ||
AK122196.1 | - | 17 - 18 | 1947 | 74 | ||
AK143907.1 | - | 17 - 18 | 1759 | 74 | ||
AK171860.1 | - | 1953 | 74 | |||
BC032216.1 | - | 8 - 9 | 778 | 74 | ||
BC141083.1 | - | 17 - 18 | 1927 | 9055695 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |