Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009509.2 | NP_033535.2 | 11 - 12 | 1263 | 69 | |
GenBank mRNA | AK027908.1 | - | 11 - 12 | 1260 | 69 | |
BC015267.1 | - | 11 - 12 | 1239 | 4236751 | 69 | |
BC117875.2 | - | 11 - 12 | 1228 | 40095718 | 69 | |
BC145671.1 | - | 11 - 12 | 1230 | 40130974 | 69 | |
M98454.1 | - | 11 - 12 | 1253 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |