Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027154.5 | NP_081430.3 | 1 - 2 | 212 | 84 | |
GenBank mRNA | AK077689.1 | - | 1 - 2 | 37 | 84 | |
AK144110.1 | - | 1 - 2 | 56 | 84 | ||
AK152864.1 | - | 1 - 2 | 55 | 84 | ||
AK166926.1 | - | 1 - 2 | 228 | 84 | ||
AK171102.1 | - | 1 - 2 | 54 | 84 | ||
BY312755.1 | - | 1 - 2 | 212 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |