Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028673.1 | NP_082949.1 | 2 - 3 | 293 | 86 | |
GenBank mRNA | AK015271.1 | - | 2 - 3 | 294 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |