Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267872.1 | NP_001254801.1 | 3 - 4 | 370 | 69 | |
GenBank mRNA | AK132808.1 | - | 3 - 4 | 371 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |