Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200004038.mm.1 |
NM_019785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004038.mm.1 |
NM_019785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004038.mm.1 |
TC1200000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004032.mm.1 |
NM_019785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004032.mm.1 |
NM_019785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004032.mm.1 |
TC1200000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004034.mm.1 |
NM_019785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004034.mm.1 |
NM_019785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004034.mm.1 |
TC1200000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004031.mm.1 |
NM_019785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004031.mm.1 |
NM_019785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004031.mm.1 |
TC1200000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004036.mm.1 |
NM_019785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004036.mm.1 |
NM_019785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004036.mm.1 |
TC1200000600.mm.1 |
MTA TC |