Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019785.2 | NP_062759.2 | 5 - 6 | 548 | 132 | |
GenBank mRNA | AF190797.1 | - | 5 - 6 | 515 | 132 | |
AK161642.1 | - | 5 - 6 | 508 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |