Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153556.1 | NP_705784.1 | 2 - 3 | 266 | 73 | |
GenBank mRNA | AK045010.1 | - | 2 - 3 | 237 | 73 | |
AK140689.1 | - | 3 - 4 | 331 | 73 | ||
BC028939.1 | - | 2 - 3 | 266 | 5364251 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |