Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133829.2 | NP_598590.2 | 6 - 7 | 2524 | 61 | |
GenBank mRNA | BC017534.1 | - | 5 - 6 | 1633 | 4488390 | 61 |
BC029324.1 | - | 5 - 6 | 1881 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |