Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290984.1 | NP_001277913.1 | 2 - 3 | 275 | 113 | |
NM_133728.3 | NP_598489.2 | 2 - 3 | 275 | 113 | ||
GenBank mRNA | AK008875.1 | - | 2 - 3 | 192 | 113 | |
AK031295.1 | - | 2 - 3 | 233 | 113 | ||
AK034746.1 | - | 193 | 113 | |||
AK049194.1 | - | 2 - 3 | 222 | 113 | ||
AK050045.1 | - | 2 - 3 | 213 | 113 | ||
AK076793.1 | - | 2 - 3 | 113 | |||
AK155030.1 | - | 2 - 3 | 214 | 113 | ||
AK168103.1 | - | 2 - 3 | 215 | 113 | ||
AK168534.1 | - | 2 - 3 | 276 | 113 | ||
AK170510.1 | - | 2 - 3 | 191 | 113 | ||
AK207754.1 | - | 2 - 3 | 102 | 113 | ||
AK214374.1 | - | 2 - 3 | 102 | 113 | ||
BC013617.1 | - | 2 - 3 | 95 | 3979452 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |