Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100002666.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002666.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002666.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002669.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002669.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002669.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002670.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002670.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002670.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002668.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002668.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002668.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002671.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002671.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002671.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002665.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002665.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002665.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002664.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002664.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002664.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002662.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002662.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002662.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002663.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002663.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002663.mm.1 |
TC0100000353.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002667.mm.1 |
NM_001160386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002667.mm.1 |
NM_001160386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002667.mm.1 |
TC0100000353.mm.1 |
MTA TC |