Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025948.2 | NP_080224.1 | 7 - 8 | 1020 | 152 | |
GenBank mRNA | AK003611.1 | - | 5 - 6 | 580 | 152 | |
AK012276.1 | - | 6 - 7 | 783 | 152 | ||
AK209661.1 | - | 1 - 2 | 148 | 152 | ||
BC031521.1 | - | 7 - 8 | 1020 | 2655161 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |