Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018868.2 | NP_061356.2 | 1 - 2 | 214 | 114 | |
GenBank mRNA | AK044216.1 | - | 211 | 114 | ||
AK133959.1 | - | 1 - 2 | 212 | 114 | ||
AK134286.1 | - | 1 - 2 | 215 | 114 | ||
BC076604.1 | - | 1 - 2 | 215 | 30630088 | 114 | |
BC085135.1 | - | 1 - 2 | 184 | 5718488 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |