Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199060.1 | NP_001185989.1 | 6 - 7 | 994 | 62 | |
NM_001199061.1 | NP_001185990.1 | 6 - 7 | 726 | 62 | ||
NM_021312.5 | NP_067287.1 | 5 - 6 | 816 | 62 | ||
GenBank mRNA | AB041608.1 | - | 6 - 7 | 995 | 62 | |
AF239765.1 | - | 5 - 6 | 472 | 62 | ||
AK007500.1 | - | 5 - 6 | 593 | 62 | ||
AK012022.1 | - | 5 - 6 | 570 | 62 | ||
AK016611.1 | - | 6 - 7 | 670 | 62 | ||
AK031436.1 | - | 5 - 6 | 579 | 62 | ||
AK149085.1 | - | 5 - 6 | 498 | 62 | ||
AK161299.1 | - | 6 - 7 | 739 | 62 | ||
AY059431.1 | - | 5 - 6 | 507 | 62 | ||
BC004748.1 | - | 5 - 6 | 650 | 3583955 | 62 | |
BC052386.1 | - | 5 - 6 | 787 | 6418498 | 62 | |
BC096651.1 | - | 5 - 6 | 561 | 1514703 | 62 | |
CX564842.1 | - | 5 - 6 | 817 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |