Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20266455 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20266458 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20266457 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
NM_001037725.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
AK078064.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
AK154812.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
BC080299.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
NM_001037725.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
AK078064.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
AK154812.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P742248 |
BC080299.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401903 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708445 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506137 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384963 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
440210 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
879261 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508635 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183115 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
228303 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
784914 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
310484 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
960080 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
255330 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
797747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360255 |
AK046396.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360255 |
AK137545.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360255 |
AK162239.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360255 |
BC023818.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360255 |
NM_198967.5 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
184453 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
474150 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
821662 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
293322 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515079 |
AK029652.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515079 |
AK031180.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515079 |
AK040613.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515079 |
AK079867.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515079 |
BC036177.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515079 |
NM_153530.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
412208 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
864268 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
585469 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
868286 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
132200 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
398739 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
278738 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916636 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
865022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760573 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
109617 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
422178 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
130869 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
230070 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
772889 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
568819 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
132084 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
964559 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
558641 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
655353 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
155137 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
144412 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94583 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
47895 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11520 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
9963 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
112843 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123922 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149698 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
NM_001037725.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
AK078064.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
AK154812.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
BC080299.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
NM_001037725.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
AK078064.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
AK154812.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P742248 |
BC080299.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P157 |
NM_001037725.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P157 |
AK013154.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P157 |
AK154812.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P157 |
BC080299.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266429 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266437 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266450 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266447 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266431 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266452 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266439 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266433 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266435 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266449 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266443 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266457 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20266454 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113887 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113896 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04114012 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113955 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113911 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123922 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113984 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113928 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149698 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04113965 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P766932 |
NM_001037725.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P766932 |
AK154812.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P766932 |
BC080299.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P157 |
NM_001037725.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P157 |
AK013154.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P157 |
AK154812.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P157 |
BC080299.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
NM_001037725.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
AK078064.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
AK154812.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
BC080299.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
NM_001037725.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
AK078064.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
AK154812.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P742248 |
BC080299.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P967353 |
NM_001037725.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P967353 |
AK154812.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P967353 |
BC080299.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04113911 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04113955 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04113984 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123922 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149698 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00601312 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20266455 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20266458 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20266457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20266457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266406 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266419 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266444 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266434 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266407 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266447 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266427 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266429 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266435 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266425 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266446 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266442 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266424 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266423 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266431 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266408 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00951138 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00087351 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266422 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00951139 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266418 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266428 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266450 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266445 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266436 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266421 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20266452 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00699351 |
- |
- |