Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027407.3 | NP_081683.3 | 7 - 8 | 1077 | 65 | |
GenBank mRNA | AK006525.1 | - | 5 - 6 | 605 | 65 | |
AK029991.1 | - | 7 - 8 | 849 | 65 | ||
AK158859.1 | - | 7 - 8 | 913 | 65 | ||
BC058983.1 | - | 2 - 3 | 206 | 65 | ||
BC079669.1 | - | 7 - 8 | 1077 | 30355976 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |