Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054066.4 | NP_473407.2 | 1 - 2 | 68 | 94 | |
GenBank mRNA | AK005949.1 | - | 1 - 2 | 74 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |