Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054066.4 | NP_473407.2 | 6 - 7 | 925 | 68 | |
GenBank mRNA | AF435950.1 | - | 6 - 7 | 930 | 68 | |
AK006672.1 | - | 6 - 7 | 970 | 68 | ||
BC106767.1 | - | 6 - 7 | 920 | 40039566 | 68 | |
BC106768.1 | - | 6 - 7 | 923 | 40039567 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |