Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177192.3 | NP_796166.2 | 13 - 14 | 3047 | 71 | |
GenBank mRNA | AK050728.1 | - | 13 - 14 | 2905 | 71 | |
AK135015.1 | - | 13 - 14 | 2930 | 71 | ||
BC132199.1 | - | 13 - 14 | 40130476 | 71 | ||
BC132535.1 | - | 13 - 14 | 2752 | 40130812 | 71 | |
BC145128.1 | - | 13 - 14 | 2768 | 71 | ||
CN537548.1 | - | 1 - 2 | 44 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |