Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023617.2 | NP_076106.2 | 4 - 5 | 438 | 63 | |
GenBank mRNA | AK054065.1 | - | 4 - 5 | 397 | 63 | |
AK143048.1 | - | 4 - 5 | 777 | 63 | ||
AK149434.1 | - | 4 - 5 | 399 | 63 | ||
BC024046.1 | - | 4 - 5 | 388 | 63 | ||
BC049853.1 | - | 4 - 5 | 390 | 63 | ||
BC138876.1 | - | 4 - 5 | 417 | 8861898 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |