Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023617.2 | NP_076106.2 | 9 - 10 | 934 | 128 | |
GenBank mRNA | AF172276.2 | - | 9 - 10 | 1010 | 128 | |
AK030109.1 | - | 5 - 6 | 473 | 129 | ||
AK149434.1 | - | 9 - 10 | 895 | 128 | ||
AY347569.1 | - | 9 - 10 | 811 | 128 | ||
AY347570.1 | - | 9 - 10 | 811 | 128 | ||
BB616838.1 | - | 5 - 6 | 473 | 128 | ||
BC049853.1 | - | 9 - 10 | 886 | 128 | ||
BC138876.1 | - | 9 - 10 | 913 | 8861898 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |