Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039710.1 | NP_001034799.1 | 1 - 2 | 226 | 152 | |
GenBank mRNA | AK002294.1 | - | 1 - 2 | 127 | 152 | |
AK012117.1 | - | 1 - 2 | 195 | 152 | ||
AK015454.1 | - | 1 - 2 | 227 | 152 | ||
AK136258.1 | - | 1 - 2 | 126 | 152 | ||
AK146260.1 | - | 1 - 2 | 120 | 152 | ||
AK168293.1 | - | 1 - 2 | 120 | 152 | ||
AK168837.1 | - | 1 - 2 | 124 | 152 | ||
BC062129.1 | - | 1 - 2 | 122 | 6506502 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |