Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172626.2 | NP_766214.2 | 6 - 7 | 1030 | 122 | |
GenBank mRNA | AY461716.1 | - | 6 - 7 | 1005 | 122 | |
BC054080.1 | - | 2 - 3 | 478 | 6390990 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |