Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
NM_001161798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
NM_001161798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
NM_010840.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
NM_010840.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
NR_027809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
NR_027809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015667.mm.1 |
TC0400001924.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015659.mm.1 |
NM_001161798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015659.mm.1 |
NM_001161798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015659.mm.1 |
NR_027809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015659.mm.1 |
NR_027809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015659.mm.1 |
TC0400001924.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
NM_001161798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
NM_001161798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
NM_010840.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
NM_010840.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
NR_027809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
NR_027809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015665.mm.1 |
TC0400001924.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015658.mm.1 |
NM_001161798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015658.mm.1 |
NM_001161798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015658.mm.1 |
NR_027809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015658.mm.1 |
NR_027809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015658.mm.1 |
TC0400001924.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400015660.mm.1 |
NM_001161798.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015660.mm.1 |
NM_001161798.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015660.mm.1 |
NR_027809.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015660.mm.1 |
NR_027809.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400015660.mm.1 |
TC0400001924.mm.1 |
MTA TC |