Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033606.2 | NP_001028778.2 | 4 - 5 | 983 | 79 | |
NM_001136222.1 | NP_001129694.1 | 3 - 4 | 565 | 79 | ||
NM_028817.3 | NP_083093.2 | 5 - 6 | 1123 | 79 | ||
GenBank mRNA | AK160839.1 | - | 3 - 4 | 509 | 79 | |
AK161708.1 | - | 5 - 6 | 1124 | 79 | ||
AK166723.1 | - | 5 - 6 | 1061 | 79 | ||
BC031529.1 | - | 3 - 4 | 708 | 4009004 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |