Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011797.2 | NP_035927.1 | 6 - 7 | 833 | 131 | |
GenBank mRNA | AB005450.1 | - | 6 - 7 | 833 | 131 | |
AK012316.1 | - | 3 - 4 | 210 | 131 | ||
AK157764.1 | - | 6 - 7 | 833 | 131 | ||
BC046995.1 | - | 6 - 7 | 815 | 6491779 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |