Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267695.2 | NP_001254624.2 | 5 - 6 | 773 | 77 | |
NM_021281.3 | NP_067256.4 | 5 - 6 | 770 | 77 | ||
GenBank mRNA | AK028366.1 | - | 5 - 6 | 772 | 77 | |
AK150274.1 | - | 5 - 6 | 704 | 77 | ||
AK150758.1 | - | 4 - 5 | 616 | 77 | ||
AK150842.1 | - | 4 - 5 | 616 | 77 | ||
AK152191.1 | - | 5 - 6 | 722 | 77 | ||
AK152618.1 | - | 4 - 5 | 616 | 77 | ||
AK153540.1 | - | 2 - 3 | 372 | 77 | ||
Y18466.1 | - | 1 - 2 | 227 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |