Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027541.4 | NP_081817.2 | 6 - 7 | 1211 | 99 | |
GenBank mRNA | AK014398.1 | - | 6 - 7 | 819 | 99 | |
AK088194.1 | - | 6 - 7 | 1212 | 99 | ||
AK139852.1 | - | 6 - 7 | 844 | 99 | ||
AK167435.1 | - | 6 - 7 | 848 | 99 | ||
BC006782.1 | - | 6 - 7 | 796 | 3589226 | 99 | |
BC026607.1 | - | 3 - 4 | 965 | 99 | ||
BC040787.1 | - | 3 - 4 | 965 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |