Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290568.1 | NP_001277497.1 | 4 - 5 | 477 | 156 | |
NM_026319.3 | NP_080595.2 | 4 - 5 | 477 | 156 | ||
GenBank mRNA | AK011690.1 | - | 4 - 5 | 448 | 156 | |
AK014770.1 | - | 4 - 5 | 445 | 156 | ||
AK053369.1 | - | 4 - 5 | 425 | 156 | ||
BC005798.1 | - | 4 - 5 | 447 | 156 | ||
BC023760.1 | - | 4 - 5 | 409 | 156 | ||
BC049156.1 | - | 4 - 5 | 397 | 156 | ||
BQ950148.1 | - | 4 - 5 | 448 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |