Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010014.3 | NP_034144.1 | 2 - 3 | 387 | 77 | |
NM_177259.4 | NP_796233.2 | 2 - 3 | 387 | 77 | ||
NR_104385.1 | 2 - 3 | 387 | 77 | |||
GenBank mRNA | BC051993.1 | - | 4 - 5 | 520 | 77 | |
BC138538.1 | - | 1 - 2 | 108 | 8861558 | 77 | |
BC138539.1 | - | 1 - 2 | 108 | 8861559 | 77 | |
BY731247.1 | - | 2 - 3 | 354 | 77 | ||
Y08379.1 | - | 2 - 3 | 335 | 77 | ||
Y08380.1 | - | 2 - 3 | 335 | 77 | ||
Y08381.1 | - | 2 - 3 | 335 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |