Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eqtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290623.1 | NP_001277552.1 | 8 - 9 | 807 | 69 | |
GenBank mRNA | AB438106.1 | - | 8 - 9 | 716 | 69 | |
AK016313.1 | - | 8 - 9 | 774 | 69 | ||
AK016738.1 | - | 8 - 9 | 778 | 69 | ||
BE632290.1 | - | 4 - 5 | 326 | 69 | ||
EU828782.1 | - | 8 - 9 | 792 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons |