Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008824.3 | NP_032850.1 | 12 - 13 | 1361 | 75 | |
GenBank mRNA | AK132629.1 | - | 12 - 13 | 1362 | 75 | |
BC132561.1 | - | 12 - 13 | 1346 | 40130838 | 75 | |
BC132563.1 | - | 12 - 13 | 1346 | 40130840 | 75 | |
BX522768.1 | - | 2 - 3 | 139 | 75 | ||
X98848.1 | - | 2 - 3 | 92 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |