Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110202.1 | NP_001103672.1 | 1 - 2 | 1275 | 83 | |
NM_001110203.1 | NP_001103673.1 | 1 - 2 | 1275 | 83 | ||
NM_001286386.1 | NP_001273315.1 | 1 - 2 | 1275 | 83 | ||
NM_001286387.1 | NP_001273316.1 | 1 - 2 | 1275 | 83 | ||
NM_001286388.1 | NP_001273317.1 | 1 - 2 | 1275 | 83 | ||
NM_053167.3 | NP_444397.2 | 1 - 2 | 1275 | 83 | ||
GenBank mRNA | AK028280.1 | - | 1 - 2 | 994 | 83 | |
AK035807.1 | - | 1 - 2 | 993 | 83 | ||
AK048875.1 | - | 1 - 2 | 1012 | 83 | ||
AK049907.1 | - | 1 - 2 | 989 | 83 | ||
AK129109.1 | - | 1 - 2 | 980 | 83 | ||
AK163123.1 | - | 1 - 2 | 1012 | 83 | ||
BC052034.1 | - | 1 - 2 | 1275 | 5684877 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |