Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164621.1 | NP_001158093.1 | 6 - 7 | 1489 | 77 | |
NM_001164622.1 | NP_001158094.1 | 4 - 5 | 1024 | 77 | ||
NM_020012.2 | NP_064396.2 | 5 - 6 | 1176 | 77 | ||
GenBank mRNA | AF249667.1 | - | 5 - 6 | 1122 | 77 | |
AF249668.1 | - | 3 - 4 | 707 | 77 | ||
AK011316.2 | - | 6 - 7 | 1177 | 77 | ||
AK038986.1 | - | 4 - 5 | 966 | 77 | ||
BC049079.1 | - | 6 - 7 | 1489 | 77 | ||
BC052205.1 | - | 3185 | 77 | |||
BC054841.1 | - | 5 - 6 | 1106 | 6489903 | 77 | |
BC094250.1 | - | 5 - 6 | 1127 | 5720753 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |