Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023645.3 | NP_076134.1 | 4 - 5 | 950 | 96 | |
GenBank mRNA | AJ404004.1 | - | 4 - 5 | 712 | 96 | |
AK007835.1 | - | 4 - 5 | 943 | 96 | ||
AK035391.1 | - | 4 - 5 | 943 | 96 | ||
AK035899.1 | - | 4 - 5 | 951 | 96 | ||
AK053315.1 | - | 4 - 5 | 943 | 96 | ||
AK079701.1 | - | 4 - 5 | 943 | 96 | ||
AK170300.1 | - | 4 - 5 | 979 | 96 | ||
AK198736.1 | - | 1 - 2 | 56 | 96 | ||
BC023141.1 | - | 4 - 5 | 930 | 4018698 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |