Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021285.3 | NP_067260.1 | 1 - 2 | 270 | 148 | |
GenBank mRNA | AK003182.1 | - | 1 - 2 | 246 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |