Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019781.2 | NP_062755.1 | 2 - 3 | 93 | 80 | |
GenBank mRNA | AF097512.1 | - | 2 - 3 | 93 | 80 | |
BC028952.1 | - | 2 - 3 | 90 | 5368312 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |