Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083342.1 | NP_001076811.1 | 13 - 14 | 2907 | 57 | |
GenBank mRNA | AB235901.1 | - | 12 - 13 | 2661 | 57 | |
AK090326.1 | - | 13 - 14 | 2928 | 57 | ||
AK173152.1 | - | 12 - 13 | 2350 | 57 | ||
BC070441.1 | - | 13 - 14 | 2781 | 30535318 | 57 | |
BC072569.1 | - | 13 - 14 | 2811 | 5719275 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |