Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113354.1 | NP_001106825.1 | 14 - 15 | 2583 | 68 | |
NM_177201.5 | NP_796175.1 | 13 - 14 | 2280 | 68 | ||
GenBank mRNA | AK036609.1 | - | 13 - 14 | 1654 | 68 | |
AK122447.1 | - | 13 - 14 | 1674 | 68 | ||
AK147939.1 | - | 14 - 15 | 1970 | 68 | ||
AK198809.1 | - | 2 - 3 | 194 | 68 | ||
BC138899.1 | - | 13 - 14 | 1549 | 8861921 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |