Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113354.1 | NP_001106825.1 | 15 - 16 | 2761 | 118 | |
NM_177201.5 | NP_796175.1 | 14 - 15 | 2458 | 118 | ||
GenBank mRNA | AK036609.1 | - | 14 - 15 | 1832 | 118 | |
AK122447.1 | - | 14 - 15 | 1852 | 118 | ||
AK147939.1 | - | 15 - 16 | 2148 | 118 | ||
BC138899.1 | - | 14 - 15 | 1727 | 8861921 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |