Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176848.1 | NP_789818.1 | 1 - 2 | 45 | 169 | |
GenBank mRNA | AK140175.1 | - | 1 - 2 | 89 | 169 | |
BC046586.1 | - | 1 - 2 | 45 | 6487759 | 169 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |