Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198292.3 | NP_938034.2 | 10 - 11 | 3239 | 112 | |
GenBank mRNA | AK129434.1 | - | 10 - 11 | 3239 | 112 | |
AK204627.1 | - | 1 - 2 | 205 | 112 | ||
BC057406.1 | - | 9 - 10 | 1643 | 5687258 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |